Skip to Content
Merck
All Photos(2)

Documents

T7189

Sigma-Aldrich

Tumor Necrosis Factor Soluble Receptor II (TNF sRII)/Fc Chimera human

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

0.004-0.016 mg per mL ED50

mol wt

52.6 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TNFRSF1B(7133)

Application

Tumor Necrosis Factor Soluble Receptor II (TNF sRII)/Fc Chimera human can be used as a test compound for analyzing the interaction between tumor necrosis factor (TNF) and its two soluble receptors by using surface plasmon resonance (SPR) technique.

Biochem/physiol Actions

Tumor Necrosis Factor Soluble Receptor II (TNF sRII)/Fc Chimera human belongs to TNF family of receptors and mediates the metabolic effects of TNF-α. Tumor Necrosis Factor Soluble Receptor II blocks the TNF-α-induced apoptosis and hence regulates the TNF-α function. It also has a pivotal role in neutralizing the biological activities of both TNF-α and TNF-β.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Analysis Note

The biological activity is measured by its ability to neutralize the TNF-α mediated cytotoxicity in mouse L-929 cells in the presence of the metabolic inhibitor actinomycin D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthews, N., et al. et al.
Lymphokines and Interferons, A Practical Approach, 221-221 (1987)
Kinetic Analysis of Interaction Between Tumor Necrosis Factor and Its Soluble Receptors.
<BIG>Fan Yang</BIG>
Journal of Analytical Chemistry, 41 (2013)
Begoña Lainez et al.
International immunology, 16(1), 169-177 (2003-12-23)
Tumor necrosis factor (TNF)-alpha is a pleiotropic cytokine involved in a broad spectrum of inflammatory and immune responses including proliferation, differentiation and cell death induction in several cell types. The biological effects of TNF-alpha are mediated via the cell-surface TNF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service