Skip to Content
Merck
All Photos(1)

Documents

T2765

Sigma-Aldrich

Thioanisole

Synonym(s):

Methyl phenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH3
CAS Number:
Molecular Weight:
124.20
Beilstein:
1904179
EC Number:
MDL number:
UNSPSC Code:
12352005

refractive index

n20/D 1.587 (lit.)

bp

188 °C (lit.)

mp

−15 °C (lit.)

density

1.057 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CSc1ccccc1

InChI

1S/C7H8S/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

HNKJADCVZUBCPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangwei Si et al.
Neuron, 101(5), 950-962 (2019-01-27)
Odor perception allows animals to distinguish odors, recognize the same odor across concentrations, and determine concentration changes. How the activity patterns of primary olfactory receptor neurons (ORNs), at the individual and population levels, facilitate distinguishing these functions remains poorly understood. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service