Skip to Content
Merck
All Photos(1)

Key Documents

SRP6487

Sigma-Aldrich

R spondin-1 human

recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE)

Synonym(s):

CRISTIN-3, CRISTIN3, FLJ40906, R-spondin-1, RP11-566C13.1, RSPO, RSPO-1, RSPO1, Rspondin, Rspondin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 27.6 kDa
observed mol wt 40 kDa by SDS-PAGE (reducing) (Ser 21 is the predicted N-terminal .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RSPO1(284654)

General description

R-spondin-1, also known as Roof plate-specific Spondin 1 (RSPO1) and cysteine rich and single thrombospondin domain containing protein 3 (Cristin 3), is a secreted protein which belongs to the R-Spondin family and encodes a secreted activator protein with two cysteine-rich, furin-like domains and one thrombospondin type 1 domain. All R spondins regulate Wnt/β-catenin signaling, but have distinct expression patterns. Like other R-Spondins, R-Spondin-1 contains two adjacent cysteine rich furin like domains (aa 34-135) with one potential N-glycosylation site, followed by a thrombospondin (TSP1) motif (aa 147-207) and a region rich in basic residues (aa 211-263). Only the furin like domains are needed for β-catenin stabilization. A putative nuclear localization signal at the C-terminus may allow some expression in the nucleus. Potential isoforms of 200 and 236 aa have an alternate, shorter N-terminus or are missing aa 146-208, respectively.

Application

R spondin-1 human has been used to study the mechanism of action of methylphenidate (a drug to treat attention-deficit hyperactivity disorder), its effect on cell proliferation and maturation.

Biochem/physiol Actions

RSPOs (roof plate-specific spondin) plays a crucial role in embryogenesis and tissue homeostasis. During the developmental stages of ovary, RSPO1 becomes a necessary factor for sex determination and follicular cell progenitor differentiation. The RSPO1 gene expression is also predicted to initiate ovarian cancer. It has a supportive role in cell proliferation, differentiation, and survival. RSPO1 is centrally associated with intestinal stem cell induction.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Methylphenidate enhances neuronal differentiation and reduces proliferation concomitant to activation of Wnt signal transduction pathways.
Grunblatt E, et al.
Translational Psychiatry, 8(1), 51-51 (2018)
RSPOs facilitated HSC activation and promoted hepatic fibrogenesis.
Yin X, et al.
Oncotarget, 7(39), 63767-63767 (2016)
Multiplicity of the interactions of Wnt proteins and their receptors.
Kikuchi A, et al.
Cellular Signalling, 19(4), 659-671 (2007)
R-Spondin1 regulates Wnt signaling by inhibiting internalization of LRP6.
Binnerts M E, et al.
Proceedings of the National Academy of Sciences of the USA, 104(37), 14700-14705 (2007)

Articles

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service