Skip to Content
Merck
All Photos(1)

Key Documents

SRP0660

Sigma-Aldrich

CD48, FC Fusion ( IgG1), AviTag (Human)

recombinant, expressed in HEK 293 cells

Synonym(s):

BLAST-1, CD2L, Cluster of Differentiation 48, SLAMF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

51 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD48(962)

General description

Human CD48, Fc fusion protein, also known as Cluster of Differentiation 48, BLAST-1, or SLAMF2. GenBank Accession No. NM_001778, a.a. 27-220 expressed in a HEK293 cell expression system. This protein is fused at the C-terminus to the Fc region of human IgG1 using Avitag technology. MW = 51 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service