Skip to Content
Merck
All Photos(1)

Documents

SML3124

Sigma-Aldrich

Gemcabene

≥98% (HPLC)

Synonym(s):

6,6′-Oxybis(2,2-dimethylhexanoic acid), CI-1027, PD72953

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O5
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C(CCCCOCCCCC(C(O)=O)(C)C)(C)C

InChI

1S/C16H30O5/c1-15(2,13(17)18)9-5-7-11-21-12-8-6-10-16(3,4)14(19)20/h5-12H2,1-4H3,(H,17,18)(H,19,20)

InChI key

SDMBRCRVFFHJKR-UHFFFAOYSA-N

Biochem/physiol Actions

Gemcabene is an orally available lipid-regulating compound that induces both a cholesterol and triglyceride reduction by a mechanism of action not linked to agonist or antagonist activity on the PPAR-α receptors. Gemcabene down-regulates apoC-III expression. Gemcabene is primarily metabolized (glucuronidation) by UDP-glucuronosyltransferase2B7 (UGT2B7). Thus, is often used as a UGT2B7 substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service