Skip to Content
Merck
All Photos(1)

Key Documents

SML1564

Sigma-Aldrich

BAY-677

≥98% (HPLC)

Synonym(s):

(S)-BAY-678, 2-Pyridinecarbonitrile, 5-[(4S)-5-acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-, 5-{(4S)-5-Acetyl-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-1,2,3,4-tetrahydropyrimidin-4-yl}pyridine-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15F3N4O2
CAS Number:
Molecular Weight:
400.35
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -21 to -29°, c = 0.5 in methanol

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(N1C2=CC=CC(C(F)(F)F)=C2)=C(C(C)=O)[C@H](C3=CN=C(C#N)C=C3)NC1=O

InChI

1S/C20H15F3N4O2/c1-11-17(12(2)28)18(13-6-7-15(9-24)25-10-13)26-19(29)27(11)16-5-3-4-14(8-16)20(21,22)23/h3-8,10,18H,1-2H3,(H,26,29)/t18-/m0/s1

InChI key

PGIVGIFOWOVINL-SFHVURJKSA-N

Biochem/physiol Actions

BAY-677 is a negative control for HNE inhibitor BAY-678.
BAY-677 is the negative control probe for BAY-678, which is a human neutrophil elastase (HNE) inhibitor with >2,000-fold selectivity for HNE over a panel of 21 other serine proteases. For characterization details of BAY-678 and BAY-677, please visit the BAY-677 probe summary on the Structural Genomics Consortium (SGC) website.

BAY-678, the active HNE probe, is available from Sigma. To learn more about and purchase BAY-678, click here.

To learn about other SGC chemical probes for protein targets, visit sigma.com/sgc

Features and Benefits

BAY-677 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service