Skip to Content
Merck
All Photos(1)

Documents

SML1469

Sigma-Aldrich

Ulipristal acetate

≥98% (HPLC)

Synonym(s):

(11β)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37NO4
CAS Number:
Molecular Weight:
475.62
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +165 to +185°, c = 1 in dichloromethane

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=C(C)[C@@]1(OC(C)=O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CCC4=C3[C@@H](C5=CC=C(N(C)C)C=C5)C[C@@]21C)=O

InChI

1S/C30H37NO4/c1-18(32)30(35-19(2)33)15-14-27-25-12-8-21-16-23(34)11-13-24(21)28(25)26(17-29(27,30)3)20-6-9-22(10-7-20)31(4)5/h6-7,9-10,16,25-27H,8,11-15,17H2,1-5H3/t25-,26+,27-,29-,30-/m0/s1

InChI key

OOLLAFOLCSJHRE-ZHAKMVSLSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ulipristal acetate is a selective progesterone receptor modulator (SPRM) with tissue-selective partial antagonist activity. It has clinical use both as an emergency contraceptive and as a treatment for uterine fibroids. Ulipristal acetate acts as a partial antagonist on the hypothalamic–pituitary–ovarian axis to inhibit or delay ovulation without affecting human embryo implantation. As a progesterone antagonist, Ulipristal acetate selectively suppresses neo-vascularization, cell proliferation, and survival in uterine fibroid cells, but not in normal myometrial cells.
Ulipristal acetate is known to reduce uncontrolled bleeding due to post uterine fibroid surgery.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulipristal acetate versus placebo for fibroid treatment before surgery.
Donnez J, et al.
The New England Journal of Medicine, 366(5), 409-420 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service