Skip to Content
Merck
All Photos(1)

Key Documents

SML1173

Sigma-Aldrich

Lubiprostone

≥98% (HPLC)

Synonym(s):

(11a,15R)-11,15-Epoxy-16,16-difluoro-15-hydroxy-9-oxo-prostan-1-oic acid, Ru 0211, Spi 0211

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32F2O5
CAS Number:
Molecular Weight:
390.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)([C@@]1(O[C@H]2[C@@H]([C@@H](C(=O)C2)CCCCCCC(=O)O)CC1)O)CCCC

InChI

1S/C20H32F2O5/c1-2-3-11-19(21,22)20(26)12-10-15-14(16(23)13-17(15)27-20)8-6-4-5-7-9-18(24)25/h14-15,17,26H,2-13H2,1H3,(H,24,25)/t14-,15+,17+,20+/m0/s1

InChI key

WGFOBBZOWHGYQH-DKYLXPRQSA-N

Biochem/physiol Actions

Lubiprostone helps to enhance intestinal permeability in humans. Hence it may inhibit leaky gut syndrome.
Lubiprostone is a bicyclic fatty acid that activates ClC-2 and CFTR chloride channels. Some reports suggest that lubiprostone opens CFTR or both CFTR and ClC-2 through interactions with the prostaglandin receptor EP4. Lubiprostone is used clinically to treat idiopathic chronic constipation and irritible bowel syndrome.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lubiprostone improves intestinal permeability in humans, a novel therapy for the leaky gut: A prospective randomized pilot study in healthy volunteers
Kato T, et al.
Testing, 12(4), e0175626-e0175626 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service