SML0624
PE 154
≥97% (HPLC)
Synonym(s):
2-(1,2,3,4-Tetrahydro-9-acridinyl)hydrazide-4-[[[7-(diethylamino)-2,4-dioxo-2H-1-benzopyran-3(4H)-ylidene]methyl]amino]-benzeneacetic acid
About This Item
Recommended Products
Assay
≥97% (HPLC)
form
powder
color
white to light brown
solubility
DMSO: 10 mg/mL, clear (warmed)
storage temp.
2-8°C
InChI
1S/C35H35N5O4/c1-3-40(4-2)24-17-18-27-31(20-24)44-35(43)28(34(27)42)21-36-23-15-13-22(14-16-23)19-32(41)38-39-33-25-9-5-7-11-29(25)37-30-12-8-6-10-26(30)33/h5,7,9,11,13-18,20-21,36H,3-4,6,8,10,12,19H2,1-2H3,(H,37,39)(H,38,41)/b28-21+
InChI key
CYZIIGRHLSZNAV-SGWCAAJKSA-N
Biochem/physiol Actions
Features and Benefits
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.
Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.
Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.
Acetylcholine is synthesized from acetyl coenzyme A and choline by the enzyme choline acetyltransferase. In addition to its synthesis in the liver, choline employed in acetylcholine production is derived from dietary sources.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service