Skip to Content
Merck
All Photos(1)

Documents

SAB4700192

Sigma-Aldrich

Monoclonal Anti-CD48 antibody produced in mouse

clone MEM-102, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BLAST-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-102, monoclonal

form

buffered aqueous solution

species reactivity

nonhuman primates, human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD48(962)

Related Categories

General description

Cluster of differentiation 48 (CD48) molecule, also known as BLAST1, is encoded by the gene mapped to human chromosome 1q23.3. CD48 is a glycosyl-phosphatidyl-inositol (GPI)-anchored cell-surface protein of immunoglobulin superfamily. The encoded protein is expressed on various hematopoietic cells.
The antibody MEM-102 reacts with CD48 antigen (Blast-1), a 40-47 kDa GPI-anchored membrane protein (immunoglobulin supergene family) widely expressed on hematopoietic cells; it is negative on granulocytes, platelets and erythrocytes.

Immunogen

Raji human Burkitt′s lymphoma cell line

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Cluster of differentiation 48 (CD48) acts as an adhesion and co-stimulatory molecule and stimulates various effects in B and T lymphocytes, natural killer cells, mast cells and eosinophils. CD48 interacts with its ligand CD2 and other molecules and promotes rearrangement of signaling factors in lipid rafts, Lck-kinase activity and tyrosine phosphorylation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the Epstein-Barr Virus-Inducible Gene Encoding the Human Leukocyte Adhesion and Activation Antigen BLAST-1 (CD48)
Fisher RC and Thorley-Lawson DA
Molecular and Cellular Biology, 11, 1614-1623 (1991)
The genetic differences between gallbladder and bile duct cancer cell lines.
Saito S
Oncology Reports, 16, 949-956 (2006)
CD48: A co-stimulatory receptor of immunity.
Elishmereni M and Levi-Schaffer F
The International Journal of Biochemistry & Cell Biology, 43, 25-28 (2011)
Soichiro Saito et al.
Oncology reports, 16(5), 949-956 (2006-10-04)
Biliary tract cancers carry dismal prognoses. It is commonly understood that chromosomal aberrations in cancer cells have prognostic and therapeutic implications. However, in biliary tract cancers the genetic changes have not yet been sufficiently studied. The aim of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service