Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101751

Sigma-Aldrich

Anti-PCSK6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PACE4, Anti-Proprotein convertase subtilisin/kexin type 6, Anti-SPC4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

91 kDa

species reactivity

human, pig, rabbit, mouse, rat, guinea pig, horse, canine, zebrafish, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PCSK6(5046)

Immunogen

The immunogen for anti-PCSK6 antibody: synthetic peptide derected towards the N terminal of human PCSK6

Sequence

Synthetic peptide located within the following region: NYDSYASYDVNGNDYDPSPRYDASNENKHGTRCAGEVAASANNSYCIVGI

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shenghan Chen et al.
Nature medicine, 21(9), 1048-1053 (2015-08-11)
Hypertension is the most common cardiovascular disease, afflicting >30% of adults. The cause of hypertension in most individuals remains unknown, suggesting that additional contributing factors have yet to be discovered. Corin is a serine protease that activates the natriuretic peptides
Ying Wang et al.
The Journal of endocrinology, 222(1), 151-160 (2014-05-27)
Mammalian proprotein convertases (PCs) play an important role in folliculogenesis, as they proteolytically activate a variety of substrates such as the transforming growth factor beta (TGFβ) superfamily. PC subtilism/kexin 6 (PCSK6) is a member of the PC family and is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service