Skip to Content
Merck
All Photos(1)

Key Documents

S7881

Sigma-Aldrich

Solanidine from potato

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO
CAS Number:
Molecular Weight:
397.64
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

~98%

storage temp.

2-8°C

SMILES string

C[C@H]1CC[C@@H]2[C@@H](C)[C@H]3[C@H](C[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]34C)N2C1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ripperger, H. and Schreiber, K.
The Alkaloids, 19, 81-81 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service