Skip to Content
Merck
All Photos(1)

Documents

P9143

Sigma-Aldrich

Phorbol 12,13-diacetate

powder

Synonym(s):

PDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O8
CAS Number:
Molecular Weight:
448.51
Beilstein:
2317312
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

color

white

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

C[C@@H]1[C@@H](OC(C)=O)[C@]2(OC(C)=O)[C@H]([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C

InChI

1S/C24H32O8/c1-11-7-17-22(29,19(11)28)9-15(10-25)8-16-18-21(5,6)24(18,32-14(4)27)20(31-13(3)26)12(2)23(16,17)30/h7-8,12,16-18,20,25,29-30H,9-10H2,1-6H3/t12-,16+,17-,18-,20-,22-,23-,24-/m1/s1

InChI key

OMHXSAMFEJVCPT-XQOWHXTBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phorbol ester with enhanced hydrophilicity.

Application

Phorbol 12,13-diacetate may be used to stimulate protein kinase C (PKC) for phosphorylation of retinal proteins.

Biochem/physiol Actions

Phorbol 12,13-diacetate (PDA) serves as a protein kinase C (PKC). It induces arrest in translocation in rod photoreceptors. PDA facilitates PKC in regulating airway contractility by inducing transient relaxation after a sustained contraction of the human isolated bronchus. PDA improves the synaptic transmission of nociceptive parabrachioamygdaloid (PBA) input onto neurons of the capsular central amygdaloid (CeAC) nucleus via activating PKC and extracellular-regulated kinase (ERK).

Other Notes

Less potent but more water soluble than phorbol 12,13-dibutyrate.

Caution

Photosensitive.

Reconstitution

Although having aqueous solubility, it is recommended that this compound first be dissolved in a water miscible organic solvent before dilution to working concentrations in aqueous media.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammadhossein Dabaghi et al.
Micromachines, 12(2) (2021-02-04)
Polydimethylsiloxane (PDMS) is a silicone-based synthetic material used in various biomedical applications due to its properties, including transparency, flexibility, permeability to gases, and ease of use. Though PDMS facilitates and assists the fabrication of complicated geometries at micro- and nano-scales
Irene Dini et al.
Antioxidants (Basel, Switzerland), 9(4) (2020-04-02)
Olive trees are grown on five continents. Fertilization of fields, pest control management, olive leaves, olive pomaces, and olive mill wastewaters have a substantial environmental impact. It is possible to reduce this problem by using organic products to cultivate and
Arrestin Translocation in Rod Photoreceptors Is Stimulated by Phorbol 12, 13--diacetate
Orisme W and Smith C
investigate opthalmology and visual science, 46, 1716-1716 (2005)
Verena Speckbacher et al.
Frontiers in microbiology, 11, 2004-2004 (2020-09-26)
The necrotrophic mycoparasite Trichoderma atroviride is a biological pest control agent frequently applied in agriculture for the protection of plants against fungal phytopathogens. One of the main secondary metabolites produced by this fungus is 6-pentyl-α-pyrone (6-PP). 6-PP is an organic
Arianna Binello et al.
Foods (Basel, Switzerland), 9(6) (2020-06-10)
Rhizomes of Curcuma longa L. are well known for their content of curcuminoids, which are compounds with interesting biological activity against various inflammatory states and diseases. Curcuminoids can degrade during processing. This piece of work investigates fast, efficient and cost-effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service