Skip to Content
Merck
All Photos(1)

Documents

P7617

Sigma-Aldrich

Prostaglandin G1

>95% (HPLC), acetone solution

Synonym(s):

15-Hydroperoxyprostaglandin R1, 8,11-Epidioxy-15-hydroperoxy-(9α,11α, 13E,15)-prost-13-ene-1-oic acid, PGG1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O6
CAS Number:
Molecular Weight:
370.48
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

>95% (HPLC)

form

acetone solution

shipped in

dry ice

storage temp.

−70°C

SMILES string

CCCCC[C@H](OO)\C=C\[C@H]1[C@H]2C[C@H](OO2)[C@@H]1CCCCCCC(O)=O

InChI

1S/C20H34O6/c1-2-3-6-9-15(24-23)12-13-17-16(18-14-19(17)26-25-18)10-7-4-5-8-11-20(21)22/h12-13,15-19,23H,2-11,14H2,1H3,(H,21,22)/b13-12+/t15-,16+,17+,18-,19+/m0/s1

InChI key

QXCRWNZYEVOQMB-CDIPTNKSSA-N

Application

Used in studies of prostacylcin synthetase

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Ham et al.
The Journal of biological chemistry, 254(7), 2191-2194 (1979-04-10)
A study of the enzymes of the arachidonic acid cascade revealed a high sensitivity of prostacyclin synthetase and a complete resistance of thromboxane A2 synthetase to time-dependent destruction by an oxidant [Ox] released during the peroxidase-catalyzed reduction of hydroperoxy fatty

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service