N8781
Nonspore anaerobic supplement
BioChemika, suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
product line
BioChemika
shelf life
limited shelf life, expiry date on the label
application(s)
microbiology
storage temp.
2-8°C
Application
An enrichment and antibiotic supplement recommended for selective isolation of nonsporing anaerobic bacteria.
Components
Ingredients: (per vial)
Sodium Pyruvate 500.00mg
Hemin 2.50mg
Nalidixic Acid 5.00mg
Menadione 0.25mg
Sodium Pyruvate 500.00mg
Hemin 2.50mg
Nalidixic Acid 5.00mg
Menadione 0.25mg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service