Skip to Content
Merck
All Photos(1)

Key Documents

N5020

Sigma-Aldrich

1-Norokadaone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H66O11
CAS Number:
Molecular Weight:
758.98
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC1CCC2(CCCCO2)OC1C(C)CC(O)C3OC4CCC5(CCC(O5)\C=C\C(C)C6CC(C)=CC7(OC(CCC7O)CC(C)=O)O6)OC4C(O)C3=C

InChI

1S/C43H66O11/c1-25-21-35(52-43(24-25)36(46)12-11-32(51-43)23-29(5)44)26(2)9-10-31-14-18-42(50-31)19-15-34-40(54-42)37(47)30(6)39(49-34)33(45)22-28(4)38-27(3)13-17-41(53-38)16-7-8-20-48-41/h9-10,24,26-28,31-40,45-47H,6-8,11-23H2,1-5H3/b10-9+

InChI key

NJOGICJCSWVJKS-MDZDMXLPSA-N

Application

May be useful as a negative control.

Other Notes

Okadaic acid analog with low protein phosphatase inhibition and little affinity for receptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nishiwaki et al.
Carcinogenesis, 11(10), 1837-1841 (1990-10-01)
Okadaic acid (OA) is a potent non-12-O-tetradecanoyl-phorbol-13-acetate (non-TPA) type tumor promoter on mouse skin. OA acts on cells through inhibiting the activity of protein phosphatases and results in the increase of phosphorylation of proteins. Seventeen OA derivatives were evaluated as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service