All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
ethanol solution
extent of labeling
60-87 Ci/mmol
shipped in
dry ice
storage temp.
−20°C
SMILES string
[H]CN1CCC[C@@H]1c2cccnc2
Gene Information
human ... CYP1A2(1544) , CYP2A6(1548)
mouse ... Cyp2a5(13087)
rat ... Chrna2(170945) , Chrnb2(54239)
Packaging
Packaged under Nitrogen
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pek Kei Im et al.
Tobacco control, 24 Suppl 4, iv40-iv47 (2015-04-19)
To determine the most prominent individual and interpersonal triggers to quit smoking in China and their associations with sociodemographic characteristics. Data come from Waves 1-3 (2006-2009) of the International Tobacco Control (ITC) China Survey, analysed cross-sectionally as person-waves (N=14,358). Measures
Association of smoking status with health-related outcomes after percutaneous coronary intervention.
Jae-Sik Jang et al.
Circulation. Cardiovascular interventions, 8(5) (2015-05-15)
Patients who smoke at the time of percutaneous coronary intervention (PCI) would ideally have a strong incentive to quit, but most do not. We sought to compare the health status outcomes of those who did and did not quit smoking
Bo Zhang et al.
American journal of epidemiology, 181(7), 513-520 (2015-03-06)
In the present study, we examined the association between duration of nicotine replacement therapy (NRT) use and smoking cessation using data from the Ontario Tobacco Survey longitudinal study (3 waves of data collected between July 2005 and December 2009). We
Xiaosi Gu et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(8), 2539-2544 (2015-01-22)
Little is known about how prior beliefs impact biophysically described processes in the presence of neuroactive drugs, which presents a profound challenge to the understanding of the mechanisms and treatments of addiction. We engineered smokers' prior beliefs about the presence
Andrew W Bergen et al.
Nicotine & tobacco research : official journal of the Society for Research on Nicotine and Tobacco, 16(12), 1638-1646 (2014-08-22)
We evaluated chr6q25.3 organic cation transporter gene (SLC22A1, SLC22A2, SLC22A3) variation and response to smoking cessation therapies. The corresponding proteins are low-affinity transporters of choline, acetylcholine and monoamines, and smoking cessation pharmacotherapies expressed in multiple tissues. We selected 7 common
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service