Skip to Content
Merck
All Photos(1)

Key Documents

M7896

Sigma-Aldrich

Diacetyl-α-Melanocyte Stimulating Hormone

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C79H111N21O20S
CAS Number:
Molecular Weight:
1706.92
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CO)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(COC(C)=O)NC(C)=O)C(=O)NC(CCC(O)=O)C(=O)NC(Cc2cnc[nH]2)C(=O)NC(Cc3ccccc3)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)NCC(=O)NC(CCCCN)C(=O)N6CCCC6C(=O)NC(C(C)C)C(N)=O

InChI

1S/C79H111N21O20S/c1-43(2)66(67(81)108)99-77(118)63-21-14-31-100(63)78(119)56(19-11-12-29-80)90-64(105)39-87-68(109)59(35-48-37-86-52-18-10-9-17-51(48)52)96-69(110)53(20-13-30-85-79(82)83)91-72(113)57(33-46-15-7-6-8-16-46)94-74(115)60(36-49-38-84-42-88-49)97-70(111)54(26-27-65(106)107)92-71(112)55(28-32-121-5)93-75(116)61(40-101)98-73(114)58(34-47-22-24-50(104)25-23-47)95-76(117)62(89-44(3)102)41-120-45(4)103/h6-10,15-18,22-25,37-38,42-43,53-63,66,86,101,104H,11-14,19-21,26-36,39-41,80H2,1-5H3,(H2,81,108)(H,84,88)(H,87,109)(H,89,102)(H,90,105)(H,91,113)(H,92,112)(H,93,116)(H,94,115)(H,95,117)(H,96,110)(H,97,111)(H,98,114)(H,99,118)(H,106,107)(H4,82,83,85)

InChI key

HVKPKJWSDNREBR-UHFFFAOYSA-N

Gene Information

human ... POMC(5443)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Lamers et al.
The Journal of endocrinology, 135(2), 285-292 (1992-11-01)
In stressed tilapia, Oreochromis mossambicus, total alpha-melanocyte-stimulating hormone (alpha-MSH) levels and di-acetyl alpha-MSH/mono-acetyl alpha-MSH (di:mono) ratios are elevated. We therefore investigated the role of alpha-MSH in the regulation of the pituitary-interrenal axis. The corticotrophic activities of des-acetyl alpha-MSH, mono-acetyl alpha-MSH
M E Goldman et al.
Endocrinology, 112(2), 435-441 (1983-02-01)
Reverse phase high performance liquid chromatography (HPLC) followed by RIA of the chromatographic fractions was used to separate and quantify, respectively, the alpha MSH-like peptides stored in the intermediate lobe (IL) of the rat pituitary gland and released from IL
M Beaulieu et al.
Endocrinology, 114(5), 1871-1884 (1984-05-01)
Treatment of rats with 2-Br-alpha-ergocryptine (bromocriptine; CB 154) elicits biochemical, physiological, and histological changes in the intermediate lobe (IL) of the rat pituitary gland, suggesting a decrease in activity in the IL. CB 154 treatment decreases 1) the capacity of
E Follenius et al.
General and comparative endocrinology, 62(2), 185-192 (1986-05-01)
The three forms of MSH(desacetyl-, monoacetyl-, and diacetyl) circulate in the carp and trout blood. Their proportions there are as follows: in the carp, the desacetylated form is 2.5% of total MSH and the monoacetylated form 72% of the acetylated
A E Lamers et al.
The American journal of physiology, 267(5 Pt 2), R1302-R1308 (1994-11-01)
After exposure of tilapia for 7 days to low-pH water, plasma thyrotropin-releasing hormone (TRH) levels were elevated, and the melanocyte-stimulating hormone (MSH) cells in the pituitary pars intermedia had increased in size and showed enhanced synthetic and secretory activity. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service