Key Documents
GE80-1106-56
Cooling tubing for MultiTemp IV thermostatic controller
L × O.D. × I.D. 4 m × 12 mm × 8 mm, pack of 1 ea
About This Item
UNSPSC Code:
41105108
NACRES:
NB.22
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Cytiva 80110656
L × O.D. × I.D.
4 m × 12 mm × 8 mm
General description
The cooling tubing, tubing connector set and hose clamps provide a flexible and safe way to connect Multiphor II to a thermostatic circulator such as MultiTemp III.
Legal Information
MultiTemp is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)