Documents
GE28-9889-65
Sigma-Aldrich
XK 50/100 Column
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
technique(s)
LPLC: suitable
General description
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to ÄKTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules.
XK empty columns provide reliable and reproducible separations. The tube material is of borosilicate glass, which allows for visual inspection of the media bed. The columns are made of biocompatible materials that exhibit excellent chemical resistance. A broad range of column dimensions makes XK columns suitable for many different purification needs. Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations. An extensive range of spare parts is available, facilitating maintenance.
Features and Benefits
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Visit the Document Library
Protocols
Column Packing and Preparation for Size Exclusion Chromatography