Skip to Content
Merck
All Photos(1)

Key Documents

G9898

Sigma-Aldrich

Gln-Ala-Thr-Val-Gly-Asp-Ile-Asn-Thr-Glu-Arg-Pro-Gly-Met-Leu-Asp-Phe-Thr-Gly-Lys

≥95% (HPLC)

Synonym(s):

Diazepam Binding Inhibitor (DBI) Fragment 51-70 human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C91H148N26O32S
CAS Number:
Molecular Weight:
2150.37
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@@H](N)CCC(N)=O)[C@@H](C)O)C(C)C)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N1CCC[C@H]1C(=O)NCC(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CCCCN)C(O)=O

Gene Information

human ... DBI(1622)

Biochem/physiol Actions

Peripheral benzodiazepine receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Cosentino et al.
Journal of leukocyte biology, 67(5), 637-643 (2000-05-16)
We studied the effects of two diazepam-binding inhibitor (DBI)-derived peptides, triakontatetraneuropeptide (DBI 17-50, TTN) and eiksoneuropeptide (DBI 51-70, ENP), on cytosolic free Ca2+ concentrations ([Ca2+]i), chemotaxis, superoxide anion (O2-) generation, and phagocytosis in human neutrophils. Both TTN and ENP induced
A S Bender et al.
Journal of neuroscience research, 54(5), 673-680 (1998-12-08)
Astroglial swelling occurs in acute hyperammonemic states, including acute hepatic encephalopathy. In these conditions, the peripheral-type benzodiazepine receptor (PBR), a receptor associated with neurosteroidogenesis, is up-regulated. This study examined the potential involvement of PBRs and neurosteroids in ammonia-induced astrocyte swelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service