Skip to Content
Merck
All Photos(1)

Key Documents

G7278

Sigma-Aldrich

Gly-Gly

BioXtra, pH 4.5-6.0 (1 M in H2O), ≥99% (titration)

Synonym(s):

Diglycine, Glycyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH2COOH
CAS Number:
Molecular Weight:
132.12
Beilstein:
1765223
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

product line

BioXtra

Assay

≥99% (titration)

impurities

Insoluble matter, passes filter test

ign. residue (900 °C)

≤0.05%

loss

≤0.05% loss on drying, 110 °C

pH

4.5-6.0 (1 M in H2O)

useful pH range

7.5-8.9

pKa (25 °C)

8.2

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
As: ≤0.00001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
NH4+: ≤0.02%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.072 at 280 in H2O at 1 M
≤0.075 at 260 in H2O at 1 M

SMILES string

NCC(=O)NCC(O)=O

InChI

1S/C4H8N2O3/c5-1-3(7)6-2-4(8)9/h1-2,5H2,(H,6,7)(H,8,9)

InChI key

YMAWOPBAYDPSLA-UHFFFAOYSA-N

Gene Information

human ... SLC15A1(6564)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P B Armentrout et al.
Journal of the American Society for Mass Spectrometry, 23(4), 632-643 (2011-09-29)
We present a full molecular description of fragmentation reactions of protonated diglycine (H(+)GG) by studying their collision-induced dissociation (CID) with Xe using a guided ion beam tandem mass spectrometer (GIBMS). Analysis of the kinetic energy-dependent CID cross sections provides the
Namrata D Udeshi et al.
Molecular & cellular proteomics : MCP, 12(3), 825-831 (2012-12-26)
Detection of endogenous ubiquitination sites by mass spectrometry has dramatically improved with the commercialization of anti-di-glycine remnant (K-ε-GG) antibodies. Here, we describe a number of improvements to the K-ε-GG enrichment workflow, including optimized antibody and peptide input requirements, antibody cross-linking
P B Armentrout et al.
Journal of the American Society for Mass Spectrometry, 23(4), 621-631 (2011-09-29)
We present a full computational description of the fragmentation reactions of protonated diglycine (H(+)GG). Relaxed potential energy surface scans performed at B3LYP/6-31 G(d) or B3LYP/6-311 + G(d,p) levels are used to map the reaction coordinate surfaces and identify the transition states (TSs) and
Daisy Bustos et al.
Molecular & cellular proteomics : MCP, 11(12), 1529-1540 (2012-06-26)
Advances in high resolution tandem mass spectrometry and peptide enrichment technologies have transformed the field of protein biochemistry by enabling analysis of end points that have traditionally been inaccessible to molecular and biochemical techniques. One field benefitting from this research
Lasse Jenner et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3812-3816 (2013-02-23)
Here we present an X-ray crystallography structure of the clinically relevant tigecycline antibiotic bound to the 70S ribosome. Our structural and biochemical analysis indicate that the enhanced potency of tigecycline results from a stacking interaction with nucleobase C1054 within the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service