Skip to Content
Merck
All Photos(1)

Key Documents

G0264

Sigma-Aldrich

D-(+)-Galactosamine hydrochloride

~99%

Synonym(s):

2-Amino-2-deoxy-D-galactopyranose hydrochloride, D-Chondrosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
3697825
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~99%

impurities

Glucosamine, essentially free

SMILES string

Cl.N[C@@H](C=O)[C@@H](O)[C@@H](O)[C@H](O)CO

InChI

1S/C6H13NO5.ClH/c7-3(1-8)5(11)6(12)4(10)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4+,5+,6-;/m0./s1

InChI key

CBOJBBMQJBVCMW-NQZVPSPJSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Similar to G 0500, but produced by Sigma.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sato et al.
Leukemia, 29(1), 145-156 (2014-05-06)
The systemic inflammatory response observed during acute graft-versus-host disease (aGVHD) is driven by proinflammatory cytokines, a 'cytokine storm'. The function of plasmin in regulating the inflammatory response is not fully understood, and its role in the development of aGVHD remains
C Hubert et al.
Journal of chromatography. A, 1395, 88-98 (2015-04-22)
When using an analytical method, defining an analytical target profile (ATP) focused on quantitative performance represents a key input, and this will drive the method development process. In this context, two case studies were selected in order to demonstrate the
E I Lafferty et al.
Genes and immunity, 15(5), 320-332 (2014-05-23)
Genetic and immunological analysis of host-pathogen interactions can reveal fundamental mechanisms of susceptibility and resistance to infection. Modeling human infectious diseases among inbred mouse strains is a proven approach but is limited by naturally occurring genetic diversity. Using N-ethyl-N-nitrosourea mutagenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service