Skip to Content
Merck
All Photos(1)

Key Documents

F8632

Sigma-Aldrich

Fmoc-Ala-OH

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-alanine, Fmoc-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33
Beilstein:
2225975
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

147-153 °C (lit.)

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m0/s1

InChI key

QWXZOFZKSQXPDC-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Sunil Kumar et al.
ACS nano, 8(5), 4157-4165 (2014-04-01)
To apply motor proteins as natural nanomolecular machines to transporting systems in nanotechnology, complete temporal control over ON/OFF switching of the motility is necessary. We have studied the photoresponsive inhibition properties of azobenzene-tethered peptides for regulation of kinesin-microtubule motility. Although
Ngoc-Duc Doan et al.
Journal of peptide science : an official publication of the European Peptide Society, 21(5), 387-391 (2014-11-18)
The solid-phase synthesis of azapeptides possessing a C-terminal aza-residue has been accomplished by a protocol featuring regioselective alkylation of benzhydrylidene-aza-glycinamide and illustrated by the syntheses of [aza-Lys(6)] growth-hormone-releasing peptide-6 analogs.
Soo Min Cho et al.
Scientific reports, 4, 6777-6777 (2014-10-28)
Amyloid-β (Aβ) is one of the few neuropathological biomarkers associated with transporters of the blood-brain barrier (BBB). Despite the well-characterized clinical indication of decreasing Aβ levels in the cerebrospinal fluid (CSF) during the development of Alzheimer's disease (AD), the link
Huiyuan Wang et al.
Molecular pharmaceutics, 11(10), 3352-3360 (2014-08-01)
Drug resistance becomes a formidable challenge against effective cancer therapy. Defective apoptosis in cancer cells is a key factor responsible for chemoresistance or radioresistance. Promoting apoptosis is an important method to sensitize the resistant cells, thereby achieving successful treatment for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service