F3380
Factor V Activating Enzyme from Russell′s viper venom
lyophilized powder, 20 units/mg protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
lyophilized powder
specific activity
20 units/mg protein
storage temp.
−20°C
Application
Factor V activating enzyme from Russell′s viper venom (RVV) is a single-chain glycoprotein that is involved in the rapid clotting of blood. Factor V circulates in the blood as an inactive cofactor and must be activated by proteases such as Factor V activating enzyme from RVV . This product may be useful in studying the blood coagulation cascade as well as the inherited deficiency called parahemophilia .
Biochem/physiol Actions
Factor V activator for RVV contains fucose, mannose, galactose, glucosamine, and neuraminic acid. Factor V activating enzyme from RVV is an arginine esterase that is sensitive to diisopropyl fluorophosphate (DFP) .
Unit Definition
One unit will increase the Factor V activity in 0.1 mL of fresh normal human plasma by at least 2× after incubation for 3 min at 37 °C.
Physical form
Stabilized in albumin and sodium chloride.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The C-terminus of tissue factor pathway inhibitor α is required for its interaction with factors V and Va.
Journal of thrombosis and haemostasis : JTH, 10(9), 1944-1946 (2012-06-29)
Acta crystallographica. Section F, Structural biology and crystallization communications, 65(Pt 12), 1306-1308 (2010-01-08)
Russell's viper venom blood coagulation factor V activator (RVV-V) is a thrombin-like serine proteinase that specifically activates factor V by cleaving a single peptide bond between Arg1545 and Ser1546. Activated factor V combines with activated factor X produced by the
The Journal of biological chemistry, 282(47), 33915-33924 (2007-09-20)
Human blood coagulation Factor V (FV) is a plasma protein with little procoagulant activity. Limited proteolysis at Arg(709), Arg(1018), and Arg(1545) by thrombin or Factor Xa (FXa) results in the generation of activated FV, which serves as a cofactor of
BMC systems biology, 6, 45-45 (2012-05-23)
Because understanding of the inventory, connectivity and dynamics of the components characterizing the process of coagulation is relatively mature, it has become an attractive target for physiochemical modeling. Such models can potentially improve the design of therapeutics. The prothrombinase complex
Snake venom proteases that activate blood-coagulation factor V.
Methods in enzymology, 80 Pt C, 275-285 (1981-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service