Skip to Content
Merck
All Photos(1)

Key Documents

F120

Sigma-Aldrich

Fura 2-AM

Synonym(s):

Fura-2 pentakis(acetoxymethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H47N3O24
CAS Number:
Molecular Weight:
1001.85
Beilstein:
8183748
MDL number:
UNSPSC Code:
12171500

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(C)cc1OCCOc2cc3cc(oc3cc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)-c4ncc(o4)C(=O)OCOC(C)=O

InChI

1S/C44H47N3O24/c1-25-7-8-32(46(16-39(53)65-20-60-26(2)48)17-40(54)66-21-61-27(3)49)35(11-25)58-9-10-59-36-12-31-13-37(43-45-15-38(71-43)44(57)69-24-64-30(6)52)70-34(31)14-33(36)47(18-41(55)67-22-62-28(4)50)19-42(56)68-23-63-29(5)51/h7-8,11-15H,9-10,16-24H2,1-6H3

InChI key

VPSRLGDRGCKUTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service