Key Documents
EP5810747007
Sigma-Aldrich
Eppendorf® Adapter for 500mL rectangular bucket
for use with 2,6-7 mL Vacutainer
About This Item
UNSPSC Code:
41121815
NACRES:
NB.02
Pricing and availability is not currently available.
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5810747007
technique(s)
centrifugation: suitable
compatibility
for use with 2,6-7 mL Vacutainer
General description
Holds 18 round-bottom tubes 2.6 – 7 mL and can also hold 500 mL rectangular beakers. To be used in Rotor A-4-81 and Rotor S-4x500. Supplied as 2 EA.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)