Documents
EP5427714006
Eppendorf® Rotor F-45-64-5-PCR
incl. rotor lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 64 x 0.2 mL PCR tubes
incl. rotor lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427714006
parameter
11,800 max. RCF (13,543 xg)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG