Skip to Content
Merck
All Photos(1)

Documents

EP0030124359

Eppendorf® PCR Eight-tube strips

capacity 0.2 mL, pkg of 960 ea (120 x strips of 8 tubes)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

colorless tube

sterility

non-sterile

feature

neck thread push fit

packaging

pkg of 960 ea (120 x strips of 8 tubes)

manufacturer/tradename

Eppendorf® 0030124359

technique(s)

PCR: suitable

capacity

0.2 mL

General description

  • Eppendorf® PCR Eight-tube strips of 0.2 mL capacity are accessories for sample handling.
  • Contamination shield on hinged lid.
  • Defined lid position due to special hinge.
  • High transparency even on the tube base.
  • Etched lid for labeling.
  • Tight sealing.
  • Easy to open.
  • For use with all thermal cyclers with 0.2 mL block format.
  • Also available in 8-tube strip format.
  • Certified free from human DNA, DNase, RNase and PCR inhibitors.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service