Skip to Content
Merck
All Photos(1)

Documents

E4404

Sigma-Aldrich

Anti-EphA8 antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Epha8(13842)

General description

The antibody demonstrates ~5% cross-reactivity with recombinant mouse EphA6, EphA7, EphA2, and EphA3 in ELISA and immunoblotting assays. Also, there is approximately 5% cross-reactivity with recombinant human EphA1 and recombinant rat EphA5.

Immunogen

purified recombinant mouse EphA8 extracellular domain.

Biochem/physiol Actions

EPH receptor A8 is a protein-tyrosine kinase family that belongs to the ephrin receptor subfamily. It regulates developmental events, particularly those of the nervous system. EphA8 binds p110γ isoform of PI3-kinase and regulates the integrin-mediated cell adhesion.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gu et al.
Molecular and cellular biology, 21(14), 4579-4597 (2001-06-21)
Recent genetic studies suggest that ephrins may function in a kinase-independent Eph receptor pathway. Here we report that expression of EphA8 in either NIH 3T3 or HEK293 cells enhanced cell adhesion to fibronectin via alpha(5)beta(1)- or beta(3) integrins. Interestingly, a
eph, the largest known family of putative growth factor receptors.
N L Tuzi et al.
British journal of cancer, 69(3), 417-421 (1994-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service