Key Documents
E0888
Horizontal submarine electrophoresis units
gel size 14 cm × 20 cm, base unit, dual mode standard gel 14 cm wide
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
Pricing and availability is not currently available.
material
acrylic
grade
electrophoresis grade
gel size
14 cm × 20 cm
General description
Submarine gels may be formed directly on the UV-transparent gel bed using small sliding baffles to contain the agarose as it cools, or outside the unit using a UV-transparent gel tray sealed with tape. Gel is 7.5 cm wide × 10 cm long. Different colors are offered for any color-coding your lab requires (e.g., RNA only).
Combs and tape are not included. Gel tray is not included with the Mini-Gel unit.
Base unit, safety cover, power leads, and all other necessary parts are included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)