D5500
1-Dehydrotestosterone acetate
Synonym(s):
1,4-Androstadien-17β-ol-3-one 17-acetate, 17β-Hydroxy-1,4-androstadien-3-one 17-acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
drug control
Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - STOT RE 2 Oral
Target Organs
Liver
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service