Skip to Content
Merck
All Photos(1)

Documents

D4516

Sigma-Aldrich

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

≥98% (TLC)

Synonym(s):

3-(N,N-Dimethyldodecylammonio)propanesulfonate, 3-(N,N-Dimethyllaurylammonio)propanesulfonate, 3-(Dodecyldimethylammonio)propanesulfonate, 3-(Lauryldimethylammonio)propanesulfonate, Dodecyldimethyl(3-sulfopropyl)ammonium hydroxide inner salt, Lauryl sulfobetaine, SB3-12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
335.55
Beilstein:
4145308
EC Number:
MDL number:
UNSPSC Code:
12161900

description

zwitterionic

Assay

≥98% (TLC)

mol wt

micellar avg mol wt 18,500

aggregation number

55

conductivity

<20 μmho per cm at 24 °C (0.1 M aqueous solution)

CMC

2-4 mM (20-25°C)

SMILES string

CCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C17H37NO3S/c1-4-5-6-7-8-9-10-11-12-13-15-18(2,3)16-14-17-22(19,20)21/h4-17H2,1-3H3

InChI key

IZWSFJTYBVKZNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate is a zwitterionic detergent that is used for protein solubilization.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidhi Puri et al.
European journal of medicinal chemistry, 45(11), 4813-4826 (2010-08-27)
In this study, we have explored the structure activity relationships of substrates of two major, promiscuous, multidrug transporters of an opportunistic human pathogen Candida albicans namely, CaCdr1p and CaMdr1p. To differentiate between substrates and non-substrates, the susceptibilities of the Saccharomyces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service