Skip to Content
Merck
All Photos(1)

Documents

D0946

Sigma-Aldrich

PDGFRα (550-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CD140A, MGC74795, Rhe-PDGFRA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

10-14 nmol/min·mg

mol wt

~95 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDGFRA(5156)

Biochem/physiol Actions

PDGFRα (platelet-derived growth factor receptor α) is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity. Aberrant expression of PDGFRα has been linked to developmental abnormalities in vertebrate models and has been implicated in multiple disease states in humans. There is widespread expression of PDGFRα in renal cell types involved in fibrotic and sclerosing processes. PDGF and its receptor PDGFRα are inducers of fibrosis in the repair phase of inflammatory bowel disease and they may also be involved in the active inflammatory phase.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Floege et al.
Journal of the American Society of Nephrology : JASN, 9(2), 211-223 (1998-04-04)
Platelet-derived growth factor (PDGF) plays an important role in renal disease. We have recently demonstrated that in healthy mature human kidney, PDGF alpha-receptor expression is largely restricted to interstitial cells. The study presented here assesses the expression of PDGF alpha-receptor
S Kumagai et al.
The Tohoku journal of experimental medicine, 195(1), 21-33 (2002-01-10)
The aim of the present study is to clarify in situ expression of platelet-derived growth factor (PDGF) and its receptors in different phases of inflammatory bowel disease (IBD). Tissues samples were obtained from 20 patients with ulcerative colitis (UC) and
Ittai Fattal et al.
Immunology, 143(3), 374-380 (2014-05-14)
Pemphigus vulgaris (PV) is an autoimmune skin disease, which has been characterized by IgG autoantibodies to desmoglein 3. Here we studied the antibody signatures of PV patients compared with healthy subjects and with patients with two other autoimmune diseases with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service