Skip to Content
Merck
All Photos(1)

Key Documents

CLS431410

Corning® glass fiber membrane filter discs

for the 150 mL funnel, diam. 43 mm

Synonym(s):

filter accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
Pricing and availability is not currently available.

Product Name

Corning® square glass fiber prefilter, for the 150 mL funnel, diam. 43 mm

material

glass fiber membrane

sterility

non-sterile

packaging

pkg of 100 ea

manufacturer/tradename

Corning 431410

diam.

43 mm

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service