Skip to Content
Merck
All Photos(1)

Documents

NCI-H69/CPR

96042328, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H69/CPR, 96042328

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human small cell lung cancer, drug-resistant

Cell Line Description

NCI-H69/CPR is a drug resistant subline of NCI-H69 (Sigma Catalogue number. 91091802). Drug resistance was developed by addition of a stepwise increase in cisplatin to the growth medium of the parental line. The cell line exhibits a 5-fold resistance to cisplatin and is cross resistant to melphalan. A significant change in cellular glutathione content or sensitivity to cadmium chloride (as indicator of metallothionein content) was not detected, but changes in glutathione-S-transferase activity were seen. Cisplatin accumulation was unchanged compared with the parent line. It is recommended to culture the cells without drugs after resuscitation until the first passage.

Application

Drug-resistance studies, small cell lung cancer

Culture Medium

RPMI 1640 + 2mM Glutamine + 0.4μg/ml cisplatin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37ºC; resuscitate in drug-free medium until first passage.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service