Skip to Content
Merck
All Photos(1)

Key Documents

OX-85

Reactivity: Anti-L-Selectin, rat, 95111405

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma spleen

ImmunoDon

BALB/c mouse spleen

Myeloma

NS1

Reactivity

Anti-L-Selectin, rat

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x Mouse BALB/c spleen

Cell Line Description

The hybridoma cell line OX-85 secretes monoclonal IgG1 (kappa) antibodies reactive with rat L-selectin.

Immunogen

Rat L-selectin

Application

Detection of rat L-selectin

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Seed at 5x10,000 cells/ml. Subculture when cells reach 5x100,000 cells/ml (usually after 3-4 days); 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service