Skip to Content
Merck
All Photos(1)

Documents

13C4

Reactivity: Anti-Shigella-like Toxin I (SLTI), 95032701

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0 Ag-14

Reactivity

Anti-Shigella-like Toxin I (SLTI)

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse Sp2/0 Ag-14 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against Shigella toxin and neutralises Shigella toxin activities from Shigella dysenteriae Type 1 and S. flexneri. It also reacts with the ß subunit of SLTI and neutralises the Shigella-like activities of E.coli, Vibrio cholerae, V.parahaemolyticus and Salmonella typhimurium.

Immunogen

Purified Shigella-like toxin from E.coli H30

Application

Study of shiga toxins.

Culture Medium

DMEM + 2mM glutamine + 0.15mg/ml oxaloacetate + 0.2U/ml Bovine Insulin + 0.05mg Sodium Pyruvate (NaP) + 5-10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service