Skip to Content
Merck
All Photos(1)

Documents

AFRC 56D2

Reactivity: Anti-Haemagglutinin, influenza, 94120826

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Horse x Mouse/Horse Hybridoma

ImmunoDon

Horse peripheral lymph node

Myeloma

AFRC 17D1

Reactivity

Anti-Haemagglutinin, influenza

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Horse peripheral lymph node x AFRC 17D1

Cell Line Description

The heterohybridoma AFRC 56D2 is one of 8 hybridoma cell lines obtained using lymphocytes from ponies immunised with influenza A equine 2 virus (isolateA/Equine/Newmarket/79 (H3N8)). For the establishment of AFRC 56D2 horse cells from the peripheral lymph node have been fused with the non-Ig-secreting heterohybridoma AFRC 17D1 (Sigma Catalogue number. 94120825).

Immunogen

Influenza A equine 2 virus

Application

Haemagglutinin detection
The isotype of the monoclonal antibody produced by this cell line is horse IgG.

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 5% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service