Skip to Content
Merck
All Photos(1)

Documents

FER

90031401, feline embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

FER, 90031401

biological source

feline embryo

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Feline endogenous retrovirus (RD 114)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Cat Domestic embryo fibroblast

Cell Line Description

The cell line produces feline endogenous retrovirus (RD 114). The cells should be handled under laboratory containment level 2.

Application

Virus studies: feline endogenous retrovirus (RD 114)

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service