Skip to Content
Merck
All Photos(1)

Key Documents

COV504

7071902, human ovary, Polygonal & fusiform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

COV504, 07071902

biological source

human ovary

packaging

tube of 5 μg 07071902-DNA-5UG
pkg of vial of cells 07071902-1VL

growth mode

Adherent

karyotype

Modal chromosome no. 64 (62-66)

morphology

Polygonal & fusiform

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human ovarian epithelial-serous carcinoma, pleural effusion

Cell Line Description

A human ovarian epithelial-serous carcinoma cell line established from a pleural effusion. Anchorage-independent growth in agar has been reported.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service