C5916
Culture tubes
size 25 mm × 150 mm, borosilicate glass
Synonym(s):
culture tube, test tube, test tubes
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
material
borosilicate glass
cap (Test tube cap)
sterility
non-sterile
packaging
case of 72 tubes
size
25 mm × 150 mm
Features and Benefits
- Kimble® culture tubes are manufactured from low extractable, chemical-resistant borosilicate glass.
- These reusable tubes feature a fire-polished top rim.
- Uniform wall thickness ensures maximum heat transfer.
- The marking spot is printed in durable white ceramic enamel.
Packaging
Packaged in a sturdy cardboard box with internal grid to minimize damage during shipping and handling.
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
recommended
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)