Skip to Content
Merck
All Photos(3)

Key Documents

C4871

Sigma-Aldrich

Anti-CTGF (181-195) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CCN2, Anti-Connective tissue growth factor precursor isoform 1, Anti-HCS24, Anti-IGFBP8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
western blot: 1:500-1:2,000

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~38 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTGF(1490)

General description

Connective tissue growth factor (CTGF) is a secreted multimodular protein, which is an important cytokine, belonging to the CCN family of multifunctional factors.

Immunogen

synthetic peptide corresponding to amino acids 181-195 of human CTGF

Application

Anti-CTGF (181-195) antibody produced in rabbit has been used in immunoblot assay to study the protein expression of cells and renal tissues. It is also suitable for western blot at a dilution of 1:500-1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Connective tissue growth factor (CTGF) is involved in several developmental regulatory processes of chondrogenesis, osteogenesis, and angiogenesis. It also has implications in various pathologcal processes, including, fibrosis, and tumorigenesis. Its gene expression is sensitive to DNA methylation. The methylation of the CTGF promoter is correlated with CTGF expression in ovarian cancer cells and may be involved in liver tumorigenesis. Its C-terminal domain plays a vital role in the S1P (sphingosine-1-phosphate)-induced endothelial cell migration.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Zhang et al.
Molecular medicine reports, 9(6), 2138-2144 (2014-03-29)
Increasing evidence shows that DNA methylation is involved in the development and progression of diabetes mellitus (DM) and its complications. Previous studies conducted by our group have indicated that high glucose levels may induce the demethylation process of the connective
Margaret Markiewicz et al.
Microcirculation (New York, N.Y. : 1994), 18(1), 1-11 (2010-12-21)
The primary objective of this study was to examine the potential interaction between S1P, a pleiotropic lipid mediator, and CTGF/CCN2, a secreted multimodular protein, in the process of endothelial cell migration. The secondary objective was to determine whether C- and
Francisco V Souza-Neto et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
We have evaluated cardiac function and fibrosis in infarcted male Wistar rats treated with MitoQ (50 mg/kg/day) or vehicle for 4 weeks. A cohort of patients admitted with a first episode of acute MI were also analyzed with cardiac magnetic
Christoph Buken et al.
Scientific reports, 9(1), 11882-11882 (2019-08-17)
The literature suggests morphological alterations and molecular biological changes within the cellular milieu of human cells, exposed to microgravity (µg), as many cell types assemble to multicellular spheroids (MCS). In this study we investigated juvenile normal human dermal fibroblasts (NHDF)
Siao-Syun Guan et al.
Scientific reports, 5, 14561-14561 (2015-10-10)
In this study, we investigated the role of ATP synthase subunit-β (ATP5b) in diabetic nephropathy. Histopathological changes, fibrosis, and protein expressions of α-smooth muscle actin (α-SMA), advanced glycation end-products (AGEs), and ATP5b were obviously observed in the kidneys of db/db

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service