Skip to Content
Merck
All Photos(1)

Key Documents

C212100

Sigma-Aldrich

DMT-2′O-Methyl-rC(tac) Phosphoramidite

Synonym(s):

DMT-2′-O-Me-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H64N5O10P
CAS Number:
Molecular Weight:
950.07
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

product line

Proligo Reagents

Quality Level

Assay

≥98.0% (reversed phase HPLC)
≥99% (31P-NMR)

form

solid

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

nucleoside profile

base: cytidine
base protecting group: TAC
2' protecting group: methyl
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1N5C=CC(NC(=O)COc6ccc(cc6)C(C)(C)C)=NC5=O

InChI

1S/C52H64N5O10P/c1-35(2)57(36(3)4)68(65-32-14-30-53)67-47-44(33-64-52(38-15-12-11-13-16-38,39-19-23-41(60-8)24-20-39)40-21-25-42(61-9)26-22-40)66-49(48(47)62-10)56-31-29-45(55-50(56)59)54-46(58)34-63-43-27-17-37(18-28-43)51(5,6)7/h11-13,15-29,31,35-36,44,47-49H,14,32-34H2,1-10H3,(H,54,55,58,59)/t44-,47-,48-,49-,68?/m1/s1

InChI key

JRJMGDLRRSEUHM-MVHRPHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service