C0478
Anti-Calpain LP-85 (Lens-Specific Calpain) (Domain IV), Large Subunit antibody produced in rabbit
~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous glycerol solution
species reactivity
mouse, rat
concentration
~1 mg/mL
technique(s)
western blot: 1:1,000
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Capn3(12335)
rat ... Capn3(29155)
Specificity
By immunoblotting against the reduced protein, the antibody reacts with bands at 85 kDa and 62 kDa and a series of further cleaved active forms. It also reacts with non-reduced calpain LP-85. The antibody does not cross-react with other calpain family members (calpain LP-82, μ-calpain, m-calpain, calpain-94, ncl-2, ncl-3, etc.).
Immunogen
synthetic peptide corresponding to a unique insert found in the ELF-hand (the calcium binding domain-IV in the large subunit) of rat LP-85 calpain (lens-specific calpain).
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service