Skip to Content
Merck
All Photos(1)

Key Documents

B9804

Sigma-Aldrich

Anti-Bcl-2 antibody, Mouse monoclonal

clone 10C4, purified from hybridoma cell culture

Synonym(s):

Anti-BCL-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10C4, monoclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

rat, mouse

concentration

~2 mg/mL

technique(s)

microarray: suitable
western blot: 2-10 μg/mL using rat osteosarcoma ROS cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Bcl2(12043)
rat ... Bcl2(24224)

Specificity

The epitope recognized by the antibody resides within amino acids 61-76 of the mouse Bcl-2 protein.

Immunogen

synthetic peptide corresponding to amino acids 61-76 of the mouse Bcl-2 sequence, conjugated to KLH.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanqi Huang et al.
Cell death & disease, 13(1), 17-17 (2021-12-22)
Impaired autophagy and excessive apoptosis disrupt cellular homeostasis and contribute to neural tube defects (NTDs), which are a group of fatal and disabling birth defects caused by the failure of neural tube closure during early embryonic development. However, the regulatory
María L Escobar et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(12), 873-889 (2019-10-05)
Oocyte cell death is a normal process in the mammalian ovary during follicular growth. Recent reports have demonstrated the presence of pro-apoptotic and pro-autophagic proteins during oocyte elimination. The goal of this study was to identify the interactions between proteins
Yue Li et al.
Molecular therapy. Nucleic acids, 35(2), 102163-102163 (2024-03-28)
Anorectal malformations (ARMs) are congenital diseases that lead to postoperative fecal incontinence, constipation, and soiling, despite improvements in surgery; however, their pathological mechanisms remain unclear. Here, we report the role of microRNA-141-3p in maintaining homeostasis between apoptosis and autophagy in
Elena Piegari et al.
Scientific reports, 10(1), 12250-12250 (2020-07-25)
Cardiotoxicity remains a serious problem in anthracycline-treated oncologic patients. Therapeutic modulation of microRNA expression is emerging as a cardioprotective approach in several cardiovascular pathologies. MiR-34a increased in animals and patients exposed to anthracyclines and is involved in cardiac repair. In
Eun-Bum Kang et al.
Apoptosis : an international journal on programmed cell death, 18(11), 1332-1347 (2013-08-03)
Alzheimer's disease (AD) is characterized by the deposition of aggregated amyloid-beta (Aβ), which triggers a cellular stress response called the unfolded protein response (UPR). The UPR signaling pathway is a cellular defense system for dealing with the accumulation of misfolded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service