Skip to Content
Merck
All Photos(2)

Documents

B2511

Sigma-Aldrich

Boc-3,4-Dehydro-L-proline

>98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Boc-3,4-Dehydro-L-proline,

Assay

>98% (TLC)

form

powder

color

white

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC=C[C@H]1C(O)=O

InChI

1S/C10H15NO4/c1-10(2,3)15-9(14)11-6-4-5-7(11)8(12)13/h4-5,7H,6H2,1-3H3,(H,12,13)/t7-/m0/s1

InChI key

BMIGSRMSSCUMAZ-ZETCQYMHSA-N

Biochem/physiol Actions

Boc-3,4-Dehydro-L-Proline is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make peptides containing 3,4-Dehydro-L-proline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Raths et al.
The Journal of biological chemistry, 263(33), 17333-17341 (1988-11-25)
alpha-Factor, a secreted tridecapeptide pheromone, is required for mating between the a- and alpha-haploid mating types of Saccharomyces cerevisiae. An analogue of alpha-factor, [DHP8,DHP11,Nle12] tridecapeptide (where DHP represents 3,4-dehydro-L-proline and Nle represents norleucine), was catalytically reduced in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service