AXYPCR2CPRTC
PCR strip tube caps
for real time PCR, thin wall, size 0.2 mL, pkg of 10x125strips/cs, clear
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
flat cap
polypropylene cap
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-2CP-RT-C
Corning
packaging
pkg of 10x125strips/cs
size
0.2 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Axygen® 1 x 8 strip flat PCR ultra-clear caps are specifically designed to fit 8 strip Real time PCR tubes.
- An even insertion cap ensures a tight seal to reduce evaporation.
- Flat cap strips feature ultra-clear upper surfaces, ideal for fluorescent, luminescent and real-time PCR.
- Light transmittance that is 75% higher than with standard strip caps.
- Manufactured with 99.9% pure virgin polypropylene.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service