Skip to Content
Merck
All Photos(1)

Key Documents

A5926

Sigma-Aldrich

N-Acetyl-D-galactosamine 4-sulfate, sodium salt, mixed anomers

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14O9NSNa
CAS Number:
Molecular Weight:
323.25
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥98%

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC(C=O)C(O)C(OS(O)(=O)=O)C(O)CO

Application

N-Acetyl-D-galactosamine 4-sulfate (GalNAc-4S) is used as a substrate to identify, differentiate and characterize N-acetylgalactosamine sulfatase(s) and arylsulfatase(s).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erno Pungor et al.
Analytical biochemistry, 395(2), 144-150 (2009-08-18)
A functional bioassay has been developed for measuring the intracellular activity of recombinant human arylsulfatase B (rhASB) on its natural glycosaminoglycan (GAG) substrates, dermatan sulfate (DS), and chondroitin sulfate (CS) when the enzyme is taken up into cultured ASB-deficient human
G J Gibson et al.
The Biochemical journal, 248(3), 755-764 (1987-12-15)
Initial purification of N-acetylgalactosamine-4-sulphate sulphatase from human liver homogenates containing approx. 1 mg of enzyme in 26 g of soluble proteins was achieved by a six-column chromatography procedure and yielded approx. 40 micrograms of a single major protein species. Enzyme
J Bielicki et al.
The Biochemical journal, 279 ( Pt 2), 515-520 (1991-10-15)
Human N-acetylgalactosamine 6-sulphatase (EC 3.1.6.14), which is involved in the lysosomal degradation of the glycosaminoglycans keratan sulphate and chondroitin 6-sulphate, was purified more than 130,000-fold in 2.8% yield from liver by an eight-step column procedure. One major form was identified
M Masue et al.
Journal of biochemistry, 110(6), 965-970 (1991-12-01)
N-Acetylgalactosamine-6-sulfate sulfatase from human placenta was purified 33,600-fold using beta-N-acetyl-D-galactosamine 6-sulfate-(1----4)-beta-D-glucuronic acid-(1----3)-N-acetyl-D-[3H]galactosaminitol 6-sulfate as the substrate. This enzyme is an oligomer with a molecular mass of 120 kDa and consists of polypeptides of 40 and 15 kDa. The 15 kDa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service