A5791
4-Androsten-4-ol-3,17-dione
Synonym(s):
4-Hydroxy-4-androstene-3,17-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
storage temp.
2-8°C
SMILES string
[H][C@@]12CCC3=C(O)C(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)C(=O)CC[C@@]24[H]
Gene Information
human ... CYP19A1(1588)
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A M Brodie et al.
Endocrinology, 100(6), 1684-1695 (1977-06-01)
4-Hydroxy-f-androstene-3,17-dione (4-OH-A) when tested at various concentrations was found to inhibit markedly the conversion of 4-andorstene-3,17-dione to estrogens inhuman placental and rat ovarian microsomes. To obtain evidence that estrogen biosynthesis could also be reduced in vivo with 4-OH-A, rats were
Ningtao Li et al.
Frontiers in endocrinology, 11, 545973-545973 (2020-10-27)
Testosterone (T), predominantly acting through its derivative 17β-estradiol (E2), regulates the brain's sexual differentiation in rodents during the perinatal sensitive period, which mirrors the window of vulnerability to the adverse effects of general anesthetics. The mechanisms of anesthesia's adverse effects
Kyoungju Choi et al.
Nanoscale research letters, 14(1), 205-205 (2019-06-19)
Gold nanoparticle (AuNP)-protein corona complexes can alter cytochrome P450 (CYP)-mediated testosterone (TST) metabolism by altering their physicochemical properties. We investigated the impact of NP size, surface chemistry, and protein corona in TST metabolism in pooled human liver microsomes (pHLM) employing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service